Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Toluenethiol 98%

Supplier: Origen Biomedical, Inc
Warning

B24970-22 B24970-14 B24970-36
AAAB24970-22EA 88.5 USD InStock
AAAB24970-22 AAAB24970-14 AAAB24970-36
4-Toluenethiol 98%
4-Toluenethiol
Purity: 98% CAS No.: 106-45-6 Molecular Formula: C7H8S Formula Weight: 124.21 Flash Point: 68°(154°F) Odor: Stench Boiling Point: 195° Beilstein Reference: 605761 Merck Reference: 14,9325 UN No.: 2811 MDL No.: MFCD00004851


Formula: C₇H₈S
Boiling Pt: 195 °C
Melting Pt: 40…44 °C
Density: 1.022 g/cm³ (51 °C)
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004851
CAS Number: 106-45-6
EINECS: 203-399-1
UN: 2811
ADR: 6.1,III
Merck Index: 12,09461
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty