H26139-MD
H26139-03
AAH26139-MDEA
126
USD
InStock
AAH26139-MD
AAH26139-03
3,5-Difluoro-4-hydroxybenzaldehyde 97%
3,5-Difluoro-4-hydroxybenzaldehyde
Purity: 97%
CAS No.: 118276-06-5
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Melting Point: 121-124°
MDL No.: MFCD06797917