Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium trifluoroacetate 98%

Supplier: Origen Biomedical, Inc
Warning

H26577-14 H26577-22
AAH26577-14EA 69.76 USD InStock
AAH26577-14 AAH26577-22
Ammonium trifluoroacetate 98%
Ammonium trifluoroacetate
Purity: 98% CAS No.: 3336-58-1 Molecular Formula: C2F3H4NO2 Formula Weight: 131.05 Melting Point: 123-126° MDL No.: MFCD03095537


Formula: C₂H₄F₃NO₂
MW: 131.05
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00012615
CAS Number: 3336-58-1
EINECS: 222-072-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty