Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Difluoromethoxy)benzaldehyde 97%

Supplier: Origen Biomedical, Inc
Warning

H26580-03 H26580-06
AAH26580-03EA 93.6 USD InStock
AAH26580-03 AAH26580-06
3-(Difluoromethoxy)benzaldehyde 97%
3-(Difluoromethoxy)benzaldehyde
Purity: 97% CAS No.: 85684-61-3 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 MDL No.: MFCD00236222


Formula: C₈H₆F₂O₂
MW: 172.13
Storage Temperature: Ambient
MDL Number: MFCD00236222
CAS Number: 85684-61-3
EINECS: None
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty