Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-6-methoxybenzoic acid 97%

Supplier: Origen Biomedical, Inc
Warning

H26830-09 H26830-03
AAH26830-09EA 375 USD InStock
AAH26830-09 AAH26830-03
2,3-Difluoro-6-methoxybenzoic acid 97%
2,3-Difluoro-6-methoxybenzoic acid
Purity: 97% CAS No.: 773873-26-0 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 126-129° MDL No.: MFCD04116018


Formula: C₈H₆F₂O₃
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD04116018
CAS Number: 773873-26-0
EINECS: None
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty