H27042-09
H27042-03
AAH27042-09EA
177
USD
InStock
AAH27042-09
AAH27042-03
1-Bromo-4-nonylbenzene 98%
1-Bromo-4-nonylbenzene
Purity: 98%
CAS No.: 51554-94-0
Molecular Formula: C15H23Br
Formula Weight: 283.25
Flash Point: 120°(248°F)
Refractive Index: 1.509
Boiling Point: 107°/0.2mm
MDL No.: MFCD00061116