Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-3-octylimidazolium tetrafluoroborate 99%

Supplier: Origen Biomedical, Inc

H27161-06 H27161-18
AAH27161-06EA 61.1 USD InStock
AAH27161-06 AAH27161-18
1-Methyl-3-octylimidazolium tetrafluoroborate 99%
1-Methyl-3-octylimidazolium tetrafluoroborate
Purity: 99% CAS No.: 244193-52-0 Molecular Formula: C12H23BF4N2 Formula Weight: 282.13 MDL No.: MFCD03095434


Formula: C₁₂H₂₃BF₄N₂
Density: 1,120
Storage Temperature: Ambient
MDL Number: MFCD03095434
CAS Number: 244193-52-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty