Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-o-toluic acid 98%

Supplier: Origen Biomedical, Inc
Danger

H27262-03 H27262-06
AAH27262-03EA 91.3 USD InStock
AAH27262-03 AAH27262-06
4-Chloro-o-toluic acid 98%
4-Chloro-o-toluic acid
Purity: 98% CAS No.: 7499-07-2 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 170-174° MDL No.: MFCD00045852


Formula: C₈H₇ClO₂
MW: 170.6
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00045852
CAS Number: 7499-07-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty