Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-2-pyridineboronic acid pinacol ester 97%

Supplier: Origen Biomedical, Inc
Warning

H27263-06 H27263-14 H27263-03
AAH27263-06EA 268 USD InStock
AAH27263-06 AAH27263-14 AAH27263-03
6-Methoxy-2-pyridineboronic acid pinacol ester 97%
6-Methoxy-2-pyridineboronic acid pinacol ester
Purity: 97% CAS No.: 1034297-69-2 Molecular Formula: C12H18BNO3 Formula Weight: 235.09 Melting Point: 68-72° MDL No.: MFCD06798266


Formula: C₁₂H₁₈BNO₃
MW: 235.09
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD06798266
CAS Number: 1034297-69-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty