Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloro-1-octanol 98%

Supplier: Origen Biomedical, Inc

H28417-AC H28417-AA
AAH28417-ACEA 133 USD InStock
AAH28417-AC AAH28417-AA
8-Chloro-1-octanol 98%
8-Chloro-1-octanol
Purity: 98% CAS No.: 23144-52-7 Molecular Formula: Cl(CH2)8OH Formula Weight: 164.67 Flash Point: 110°(230°F) Refractive Index: 1.458 Boiling Point: 129-130/11mm MDL No.: MFCD00040005


Formula: C₈H₁₇ClO
MW: 164.68
Boiling Pt: 233
Melting Pt: -3
Density: 0.976
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040005
CAS Number: 23144-52-7
EINECS: 245-451-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty