Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-methylbutanone tech. 85%, Technical Grade

Supplier: Origen Biomedical, Inc
Warning

H28483-06 H28483-03
AAH28483-06EA 501 USD InStock
AAH28483-06 AAH28483-MD AAH28483-03
4-Hydroxy-3-methylbutanone tech. 85%, Technical Grade
4-Hydroxy-3-methylbutanone
Purity: 85% CAS No.: 3393-64-4 Molecular Formula: C5H10O2 Formula Weight: 102.13 Refractive Index: 1.4325


Formula: C₅H₁₀O₂
Boiling Pt: 90 °C (1013 hPa)
Density: 0.9908 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004739
CAS Number: 3393-64-4
EINECS: 222-238-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty