Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amantadine 98%

Supplier: Origen Biomedical, Inc
Warning

H30076-22 H30076-06 H30076-14
AAH30076-22EA 526.74 USD InStock
AAH30076-22 AAH30076-06 AAH30076-14
Amantadine 98%
Amantadine
Purity: 98% CAS No.: 768-94-5 Molecular Formula: C10H17N Formula Weight: 151.25 Melting Point: ca 200° subl. MDL No.: MFCD00074732


Formula: C₁₀H₁₇N
MW: 151.25 g/mol
Boiling Pt: 214
Melting Pt: 82 (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00074732
CAS Number: 768-94-5
EINECS: 212-201-2
Merck Index: 12,00389
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty