Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminothiobenzamide 97%

Supplier: Origen Biomedical, Inc
Warning

H51809-03 H51809-06
AAH51809-03EA 46.2 USD InStock
AAH51809-03 AAH51809-06
2-Aminothiobenzamide 97%
2-Aminothiobenzamide
Purity: 97% CAS No.: 2454-39-9 Molecular Formula: C7H8N2S Formula Weight: 152.21 Melting Point: 115-120° MDL No.: MFCD00963496


Formula: C₇H₈N₂S
MW: 152.22
Melting Pt: 115…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00963496
CAS Number: 2454-39-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty