L01293-06
L01293-22
L01293-14
AAAL01293-06EA
40.9
USD
InStock
AAAL01293-06
AAAL01293-22
AAAL01293-14
2-Methoxybenzonitrile 98%
2-Methoxybenzonitrile
Purity: 98%
CAS No.: 6609-56-9
Molecular Formula: C8H7NO
Formula Weight: 133.15
Flash Point: 110°(230°F)
Refractive Index: 1.5465
Odor: Almond-like
Boiling Point: 256°
Beilstein Reference: 2207134
UN No.: 3276
MDL No.: MFCD00001783