L02637-AU
L02637-AK
AAAL02637-AUEA
144
USD
InStock
AAAL02637-AU
AAAL02637-AK
1-Hexadecene 94%
1-Hexadecene
Purity: 94%
CAS No.: 629-73-2
Molecular Formula: CH3(CH2)13CH=CH2
Formula Weight: 224.43
Flash Point: 132°(269°F)
Refractive Index: 1.4420
Melting Point: 4°
Boiling Point: 273-274°
Beilstein Reference: 1753401
MDL No.: MFCD00008991