L03083-22
L03083-14
AAAL03083-22EA
67.89
USD
InStock
AAAL03083-22
AAAL03083-14
1,2-Dichloropropane 98%
1,2-Dichloropropane
Purity: 98%
CAS No.: 78-87-5
Molecular Formula: C3H6Cl2
Formula Weight: 112.99
Flash Point: 4°(39°F)
Refractive Index: 1.4380
Odor: Sweet
Melting Point: -100°
Boiling Point: 95-97°
Beilstein Reference: 1718880
Merck Reference: 14,7854
UN No.: 1279
MDL No.: MFCD00000868