Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorotributylsilane 97%

Supplier: Origen Biomedical, Inc
Danger

L03477-14 L03477-06
AAAL03477-14EA 204 USD InStock
AAAL03477-14 AAAL03477-06
Chlorotributylsilane 97%
Chlorotributylsilane
Purity: 97% CAS No.: 995-45-9 Molecular Formula: C12H27ClSi Formula Weight: 234.89 Flash Point: 104°(219°F) Refractive Index: 1.4460 Boiling Point: 93-94°/4mm Beilstein Reference: 1742537 UN No.: 2987 MDL No.: MFCD00000509


Formula: C₁₂H₂₇ClSi
Boiling Pt: 93…94 °C/4 mm
Density: 0.878 g/cm³ (20 °C)
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000509
CAS Number: 995-45-9
EINECS: 213-627-1
UN: 2987
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty