L03703-06
L03703-14
AAAL03703-06EA
49.41
USD
InStock
AAAL03703-06
AAL03703-14
Allyltrichlorosilane 95%
Allyltrichlorosilane
Purity: 95%
CAS No.: 107-37-9
Molecular Formula: C3H5Cl3Si
Formula Weight: 175.52
Flash Point: 18°(64°F)
Refractive Index: 1.4490
Boiling Point: 117-118°
Beilstein Reference: 1743449
UN No.: 1724
MDL No.: MFCD00000486