Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentamethylbenzene 99%

Supplier: Origen Biomedical, Inc
Warning

L04163-06 L04163-22 L04163-14
AAAL04163-06EA 40.1 USD InStock
AAAL04163-06 AAAL04163-22 AAL04163-14
Pentamethylbenzene 99%
Pentamethylbenzene
Purity: 99% CAS No.: 700-12-9 Molecular Formula: C11H16 Formula Weight: 148.25 Flash Point: 91°(195°F) Melting Point: 50-51° Boiling Point: 231-232° Beilstein Reference: 1905349 UN No.: 1325 MDL No.: MFCD00008522


Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 232
Melting Pt: 50…51 °C
Density: 0.917
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008522
CAS Number: 700-12-9
EINECS: 211-837-8
UN: 1325
ADR: 4.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty