L04372-14
L04372-06
AAAL04372-14EA
217
USD
InStock
AAAL04372-14
AAAL04372-06
N-Methylethylamine 98+%
N-Methylethylamine
Purity: 98+%
CAS No.: 624-78-2
Molecular Formula: CH3CH2NHCH3
Formula Weight: 59.11
Storage Temperature: 2°C to 8°C
Flash Point: -34°(-29°F)
Refractive Index: 1.3740
Odor: Amine-like
Boiling Point: 33-34°
Beilstein Reference: 1730786
UN No.: 2733
MDL No.: MFCD00009030