Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hexadecanone 95%

Supplier: Origen Biomedical, Inc

L05098-06 L05098-14
AAAL05098-06EA 47.1 USD InStock
AAAL05098-06 AAAL05098-14
3-Hexadecanone 95%
3-Hexadecanone
Purity: 95% CAS No.: 18787-64-9 Molecular Formula: C16H32O Formula Weight: 240.43 Melting Point: 41-45° Boiling Point: 140°/2mm MDL No.: MFCD00048458


Formula: C₁₆H₃₂O
Boiling Pt: 140 °C/2 mm
Melting Pt: 41…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00048458
CAS Number: 18787-64-9
EINECS: 242-572-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty