L07033-06
L07033-14
AAAL07033-06EA
94.1
USD
InStock
AAAL07033-06
AAAL07033-14
7,8-Epoxyoctene 97%
7,8-Epoxyoctene
Purity: 97%
CAS No.: 19600-63-6
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 38°(100°F)
Refractive Index: 1.4350
Boiling Point: 167°
Beilstein Reference: 1305290
UN No.: 1993
MDL No.: MFCD00005156