L07981-06
L07981-14
AAAL07981-06EA
66.6
USD
InStock
AAAL07981-06
AAAL07981-14
2-Chlorobenzoxazole 99%
2-Chlorobenzoxazole
Purity: 99%
CAS No.: 615-18-9
Molecular Formula: C7H4ClNO
Formula Weight: 153.57
Storage Temperature: 2°C to 8°C
Flash Point: 85°(185°F)
Refractive Index: 1.5660
Boiling Point: 201-202°
Beilstein Reference: 115982
MDL No.: MFCD00005766