Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-α-Methoxyphenylacetic acid 99%

Supplier: Origen Biomedical, Inc
Warning

L08719-03 L08719-MD
AAAL08719-03EA 116 USD InStock
AAAL08719-03 AAAL08719-MD
(R)-(-)-α-Methoxyphenylacetic acid 99%
(R)-(-)-α-Methoxyphenylacetic acid
Purity: 99% CAS No.: 3966-32-3 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 63-68° Boiling Point: 164-166°/18mm MDL No.: MFCD00004250


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 164…166 °C (18 mmHg)
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004250
CAS Number: 3966-32-3
EINECS: 223-580-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty