L09464-03
L09464-06
AAAL09464-03EA
96.4
USD
InStock
AAAL09464-03
AAAL09464-06
(R)-(-)-2-Phenylbutyric acid 99%
(R)-(-)-2-Phenylbutyric acid
Purity: 99%
CAS No.: 938-79-4
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: 110°(230°F)
Refractive Index: 1.5150
Boiling Point: 270-272°
Beilstein Reference: 2327346
Optical Rotation: -100° (c=5 in toluene)
MDL No.: MFCD00063165