Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triisopropylsilane 98%

Supplier: Origen Biomedical, Inc
Warning

L09585-06 L09585-14 L09585-22
AAAL09585-06EA 34.36 USD InStock
AAAL09585-06 AAAL09585-14 AAAL09585-22
Triisopropylsilane 98%
Triisopropylsilane
Purity: 98% CAS No.: 6485-79-6 Molecular Formula: C9H22Si Formula Weight: 158.36 Flash Point: 35°(95°F) Refractive Index: 1.4340 Odor: Mild Boiling Point: 166° Beilstein Reference: 1733718 UN No.: 1993 MDL No.: MFCD00009657


Formula: C₉H₂₂Si
MW: 158.36 g/mol
Boiling Pt: 84
Density: 0.773
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009657
CAS Number: 6485-79-6
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty