L09970-14
L09970-06
L09970-03
AAAL09970-14EA
413
USD
InStock
AAAL09970-14
AAAL09970-06
AAAL09970-03
Thiazole 99%
Thiazole
Purity: 99%
CAS No.: 288-47-1
Molecular Formula: C3H3NS
Formula Weight: 85.13
Flash Point: 22°(71°F)
Refractive Index: 1.5390
Odor: Unpleasant
Melting Point: -33°
Boiling Point: 117-118°
Beilstein Reference: 103852
Merck Reference: 14,9307
UN No.: 1993
MDL No.: MFCD00005315