L10697-06
L10697-03
AAAL10697-06EA
227.99
USD
InStock
AAAL10697-06
AAAL10697-03
3,5-Dichlorothiophenol 97%
3,5-Dichlorothiophenol
Purity: 97%
CAS No.: 17231-94-6
Molecular Formula: C6H4Cl2S
Formula Weight: 179.07
Odor: Unpleasant
Melting Point: 60-63°
Boiling Point: 270-272°
Beilstein Reference: 2041184
MDL No.: MFCD00041423