Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trimethoxybenzylamine 96%

Supplier: Origen Biomedical, Inc
Danger

L13461-06 L13461-14
AAAL13461-06EA 72.8 USD InStock
AAAL13461-06 AAAL13461-14
3,4,5-Trimethoxybenzylamine 96%
3,4,5-Trimethoxybenzylamine
Purity: 96% CAS No.: 18638-99-8 Molecular Formula: C10H15NO3 Formula Weight: 197.24 Flash Point: 110°(230°F) Refractive Index: 1.5480 Boiling Point: 120-122°/0.05m Beilstein Reference: 1107570 UN No.: 2735 MDL No.: MFCD00017150


Formula: C₁₀H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00017150
CAS Number: 18638-99-8
UN: 2735
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty