L14521-AC
L14521-AE
AAAL14521-ACEA
31.05
USD
InStock
AAAL14521-AC
AAAL14521-AE
N,N-Dimethylcyclohexylamine 98+%
N,N-Dimethylcyclohexylamine
Purity: 98+%
CAS No.: 98-94-2
Molecular Formula: C8H17N
Formula Weight: 127.23
Flash Point: 39°(102°F)
Refractive Index: 1.4540
Melting Point: -60°
Boiling Point: 160-161°
Beilstein Reference: 1919922
UN No.: 2264
MDL No.: MFCD00003844