Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Ethylindole 98+%

Supplier: Origen Biomedical, Inc

L14875-09 L14875-18
AAAL14875-09EA 67.8 USD InStock
AAAL14875-09 AAAL14875-18
7-Ethylindole 98+%
7-Ethylindole
Purity: 98+% CAS No.: 22867-74-9 Molecular Formula: C10H11N Formula Weight: 145.21 Refractive Index: 1.6030 Boiling Point: 230° MDL No.: MFCD00143514


Formula: C₁₀H₁₁N
MW: 145.2 g/mol
Boiling Pt: 230
Density: 1.058
Storage Temperature: Ambient
MDL Number: MFCD00143514
CAS Number: 22867-74-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty