L15008-36
L15008-22
AAAL15008-36EA
126
USD
InStock
AAAL15008-36
AAAL15008-22
Hydroxyacetone 95%
Hydroxyacetone
Purity: 95%
CAS No.: 116-09-6
Molecular Formula: C3H6O2
Formula Weight: 74.08
Storage Temperature: 2°C to 8°C
Flash Point: 56°(132°F)
Refractive Index: 1.4250
Odor: Characteristic
pH: 3.4
Melting Point: -17°
Boiling Point: 145-146°
Beilstein Reference: 605368
Merck Reference: 14,65
UN No.: 1224
MDL No.: MFCD00004669