Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexyltriethoxysilane 97%

Supplier: Origen Biomedical, Inc
Warning

L16521-22 L16521-14
AAAL16521-22EA 115 USD InStock
AAAL16521-22 AAAL16521-14
Hexyltriethoxysilane 97%
Hexyltriethoxysilane
Purity: 97% CAS No.: 18166-37-5 Molecular Formula: C12H28O3Si Formula Weight: 248.44 Flash Point: 95°(203°F) Boiling Point: 115°/18mm MDL No.: MFCD00087744


Formula: C₁₂H₂₈O₃Si
Boiling Pt: 115 °C/18 mm
Density: 0.860
Flash Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00087744
CAS Number: 18166-37-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty