L19235-14
L19235-06
AAAL19235-14EA
522.99
USD
InStock
AAAL19235-14
AAAL19235-03
AAAL19235-06
(4-Fluorophenyl)acetyl chloride 97%
(4-Fluorophenyl)acetyl chloride
Purity: 97%
CAS No.: 459-04-1
Molecular Formula: C8H6ClFO
Formula Weight: 172.59
Flash Point: 107°(224°F)
Refractive Index: 1.5120
Boiling Point: 60°/8mm
Beilstein Reference: 972148
UN No.: 3265
MDL No.: MFCD00674181