Z-Competent™ Transformation, Escherichia coli, G-Biosciences
Supplier: G-Biosciences
GZ-4 GZ-5
82023-074EA 379.39 USD InStock
82023-074 82023-076
Z-Competent™ Transformation, Escherichia coli, G-Biosciences
Competent Cells
Z-Competent™ E
The Z-Competent™ E. coli Transformation kit features a transformation efficiency of 2 x 10⁸ – 1 x 10⁹ transformants per microgram supercoiled pUC19 plasmid DNA. The efficiency of transformation varies depending on E. coli strain and plasmid size. The competent cells can be used immediately or stored for later use.
Other features of G-Biosciences' Z-Competent™ E. coli Transformation kit include: no heat shock requirement, transformations in under five minutes, and competent cells in less than 20 minutes.
G-Biosciences offers two different kits: one is supplied with enough reagent sufficient for processing up to 300 preps; the other kit is supplied with reagents as well as SOB Culture Medium and is sufficient for processing up to one hundred preps (100 microliter aliquots of competent cells).
coli Transformation kit is a new simple method designed to rapidly generate competent E. coli cells for highly efficient E. coli transformation. E. coli are grown in SOB medium, then washed and suspended in our supplied Competent Buffer. The bacterial cells are then ready for transformation.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty