Precoiled Pipe Markers- 'Condensate Drain', Black/Yellow, National Marker
Supplier: National Marker
A4025 B4025 C4025 D4025 E4025 F4025 G4025 H4025
94003-330EA 17.79 USD InStock
94003-330 94004-104 94005-500 94009-412 94012-592 94013-788 94014-334 94015-114
Precoiled Pipe Markers- 'Condensate Drain', Black/Yellow, National Marker
Signs
Use on wet, oily and dusty pipes where pressure sensitive vinyl markers won't hold
Can be removed and reapplied in other areas.
Certificaciones: Meets ANSI A13.1-2007.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty