Inspection Mirrors, Ball Joint, Excelta Corp®
Supplier: Excelta
415 416 417
63042-178EA 32.19 USD InStock
63042-178 63042-180 63042-182
Inspection Mirrors, Ball Joint, Excelta Corp®
Mirrors
Inspection mirrors are critical in providing visibility in restricted areas, with a unique ball joint to position mirrors for the most difficult applications.
  • Ball-jointed allows for unlimited positioning
  • Vinyl grips for comfort
These three star round/oval inspection mirrors have insulated vinyl grips. The clear-view mirrors are copper backed and electro-plated. They feature a 'ball-joing' for positioning the mirror in the perfect position.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty