Adapter, "Midi" #11 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5261-37 5261-35 5261-36 5261-39 5261-38 5261-40
89051-490EA 101.72 USD InStock
89051-490 89051-494 89051-496 89051-502 89051-508 89051-512
Adapter, "Midi" #11 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This "midi" adapter has a standard taper inner joint at the bottom, #11 Ace-Thred at the top and a serrated hose connection.
  • Ace-Thred at top
  • Bottom [ST] inner joint
  • Serrated hose connection
  • Fabricated from borosilicate glass
Adapter will accept inner tubes with diameters of 9mm to 10.5mm such as thermowells, gas dispersion tubes, and vacuum takeoffs. Suitable for most vacuum work.
Información para pedidos: Available complete with a 89059-214 nylon bushing and 89060-966 FETFE o-ring, or as glassware only.
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty