Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Brij® C20, Brij® for use in aqueous, textile, chemical and industrial foam control

Supplier: SPECTRUM CHEMICAL MFG CORP

BR106-12KG BR106-25GM BR106-2.5KG
700012-472EA 1261.4 USD InStock
700012-472 700012-476 700012-478 700012-474
Brij® C20, Brij® for use in aqueous, textile, chemical and industrial foam control
Brij® C20
Brij(R) C20 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.


Formula: C₁₆H₃₃(OCH₂CH₂)₂₀OH
MW: 1123.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080892
CAS Number: 9004-95-9
EINECS: 500-014-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty