Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyvinyl alcohol 99-100%, Reagent Grade, hydrolysed

Supplier: SPECTRUM CHEMICAL MFG CORP

PO141-1KG PO141-250GM
700012-246EA 316.05 USD InStock
700012-246 700012-248
Polyvinyl alcohol 99-100%, Reagent Grade, hydrolysed
Polyvinyl alcohol
Polyvinyl Alcohol, 99-100 Percent, Hydrolyzed, Reagent, also known as PVA or PVAI, is a synthetic polymer used in textiles, papermaking, and several types of coatings. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.


Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty