Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ytterbium(III) fluoride 99 for synthesis

Supplier: MATRIX SCIENTIFIC MS

001026-5G 001026-25G
101780-874EA 35.38 USD InStock
101780-874 101780-876
Ytterbium(III) fluoride 99 for synthesis
Ytterbium(III) fluoride
Hazard code(s): Toxic


Formula: F₃Yb
MW: 230.04
Boiling Pt: 2227
Melting Pt: 1163.85…1179.85 °C
Density: 8.2 g/cm³ (20 °C)
MDL Number: MFCD00049615
CAS Number: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 1157°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty