Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)anthranilic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

001367-5G 001367-25G
101781-504EA 35.38 USD InStock
101781-504 101781-506
3-(Trifluoromethyl)anthranilic acid ≥98% for synthesis
3-(Trifluoromethyl)anthranilic acid
Hazard code(s): Irritant


Formula: C₈H₆F₃NO₂
MW: 205.14
Melting Pt: 157…160 °C
MDL Number: MFCD00274210
CAS Number: 313-12-2
Order Now
Specification Test Results
Melting point 157-160°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty