Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-(trifluoromethoxy)benzene ≥99% for synthesis

Supplier: MATRIX SCIENTIFIC MS

001378-25G 001378-100G
101781-526EA 36.92 USD InStock
101781-526 101781-528
1-Bromo-2-(trifluoromethoxy)benzene ≥99% for synthesis
1-Bromo-2-(trifluoromethoxy)benzene
Hazard code(s): Irritant


Formula: C₇H₄BrF₃O
Boiling Pt: 54…56 °C (15mm)
MDL Number: MFCD00040942
CAS Number: 64115-88-4
Order Now
Specification Test Results
Boiling point 54-56°C/15m

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty