Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)-p-anisidine ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

001444-5G 001444-25G
101781-670EA 29.23 USD InStock
101781-670 101781-672
3-(Trifluoromethyl)-p-anisidine ≥98% for synthesis
3-(Trifluoromethyl)-p-anisidine
Hazard code(s): Irritant


Formula: C₈H₈F₃NO
MW: 191.15
Boiling Pt: 112
Melting Pt: 58…59 °C
MDL Number: MFCD00043460
CAS Number: 393-15-7
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Boiling point 112°C/18mm
Melting point 58-59°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty