Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-3-(trifluoromethyl)benzoic acid for synthesis

Supplier: MATRIX SCIENTIFIC MS

001446-1G 001446-5G
101781-678EA 38.46 USD InStock
101781-678 101781-680
4-Methoxy-3-(trifluoromethyl)benzoic acid for synthesis
4-Methoxy-3-(trifluoromethyl)benzoic acid
Hazard code(s): Irritant


Formula: C₉H₇F₃O₃
MW: 220.15
Melting Pt: 169…171 °C
MDL Number: MFCD01091014
CAS Number: 213598-09-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty