Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-(trifluoromethyl)anisole for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

001451-5G 001451-25G
101781-682EA 41.54 USD InStock
101781-682 101781-684
4-Bromo-2-(trifluoromethyl)anisole for synthesis
4-Bromo-2-(trifluoromethyl)anisole
Hazard code(s): Irritant


Formula: C₈H₆BrF₃O
MW: 255.03
Boiling Pt: 120 °C (30mm)
MDL Number: MFCD01631579
CAS Number: 1514-11-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty