Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thiopheneboronic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

001453-5G 001453-25G
101781-692EA 56.92 USD InStock
101781-692 101781-694
3-Thiopheneboronic acid ≥98% for synthesis
3-Thiopheneboronic acid
Hazard code(s): Irritant, Irritant-Harmful, Store in Freezer


Formula: C₄H₅BO₂S
MW: 127.96
Melting Pt: 169…170 °C
MDL Number: MFCD00151851
CAS Number: 6165-69-1
Order Now
Specification Test Results
Melting point 169-170°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty