Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,11H-Eicosafluoro-1-undecanol ≥85%, tech.

Supplier: MATRIX SCIENTIFIC MS

001536-25G 001536-100G 001536-500G
101781-920EA 52.31 USD InStock
101781-920 101781-922 101781-924
1H,1H,11H-Eicosafluoro-1-undecanol ≥85%, tech.
1H,1H,11H-Eicosafluoro-1-undecanol
Hazard code(s): Irritant


Formula: C₁₁H₄F₂₀O
Boiling Pt: 180…181 °C (200mm)
Melting Pt: 95…97 °C
MDL Number: MFCD00039628
CAS Number: 307-70-0
EINECS: 206-209-5
Order Now
Specification Test Results
Boiling point 180-181°C/2
Melting point 95-97°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty