Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chlorotoluene ≥98%

Supplier: MATRIX SCIENTIFIC MS

001657-5G 001657-25G
101782-212EA 23.08 USD InStock
101782-212 101782-214
5-Bromo-2-chlorotoluene ≥98%
5-Bromo-2-chlorotoluene
Hazard code(s): Irritant


Formula: C₇H₆BrCl
Boiling Pt: 115…117 °C (20mm)
Density: 1.53 g/cm³
MDL Number: MFCD00018503
CAS Number: 54932-72-8
Order Now
Specification Test Results
Boiling point 115-117°C/2
Density 1.53

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty