Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoroanthranilic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

001693-25G 001693-100G
101782-294EA 36.92 USD InStock
101782-294 101782-296
5-Fluoroanthranilic acid ≥97% for synthesis
5-Fluoroanthranilic acid
Hazard code(s): Irritant


Formula: C₇H₆FNO₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00055566
CAS Number: 446-08-2
EINECS: 207-159-7
Order Now
Specification Test Results
Melting point 182-184°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty